Merck
Joint-action of antifouling substances in copper-free paints.
Colloids and surfaces. B, Biointerfaces (2012-10-30)
Fabienne Faÿ, David Carteau, Isabelle Linossier, Maxime Delbury, Karine Vallée-Réhel
ABSTRACT
Due to the environmentally harmful impact of tributyltin self-polishing paints, there is a critical need of more ecological alternatives. The aim of the present work is to study the joint-action of three molecules chosen in order to combine the two modes of prevention: chemical and physical repelling of biofouling. This "hybrid" system is principally dedicated to disturb durable settlement of microfouling. Each component was chosen according to its specific properties: chlorhexidine is a bisdiguanide antiseptic with antibacterial activity, zinc peroxide is an inorganic precursor of high instable entities which react with seawater to create hydrogen peroxide, Tween 85 is a non ionic surfactant disturbing interactions between colonizing organisms and surface. Obtained results highlighted the interest on mixing such molecules to get additive action on antifouling efficiency.
Sigma-Aldrich
TWEEN® 85
Pricing and availability is not currently available.
Sigma-Aldrich
ECO TWEEN® 85
Pricing and availability is not currently available.