Merck
  • Evaluation of three experimental bovine viral diarrhea virus killed vaccines adjuvanted with combinations of Quil A cholesterol and dimethyldioctadecylammonium (DDA) bromide.
Evaluation of three experimental bovine viral diarrhea virus killed vaccines adjuvanted with combinations of Quil A cholesterol and dimethyldioctadecylammonium (DDA) bromide.
Veterinary research communications (2010-09-28)
Julia F Ridpath, Paul Dominowski, Ramasany Mannan, Robert Yancey, James A Jackson, Lucas Taylor, Sangita Mediratta, Robert Eversole, Charles D Mackenzie, John D Neill
ABSTRACT
Bovine viral diarrhea virus (BVDV) infections cause respiratory, reproductive, and enteric disease in cattle. Vaccination raises herd resistance and limits the spread of BVDV among cattle. Both killed and modified live vaccines against BVDV are available. While modified live vaccines elicit an immune response with a broader range and a longer duration of immunity, killed vaccines are considered to be safer. One way to improve the performance of killed vaccines is to develop new adjuvants. The goal of this research was evaluate new adjuvants, consisting of combinations of Quil A cholesterol and dimethyldioctadecylammonium (DDA) bromide, for use in killed vaccines. Responses to three novel killed vaccines, using combinations of Quil A and DDA as adjuvants, were compared to responses to a commercial modified live and a commercial killed vaccine. Vaccination response was monitored by measuring viral neutralizing antibodies (VN) levels and by response to challenge. All three novel vaccines were efficacious based on reduction in virus isolation, pyrexia, and depression. Compared to a commercial killed vaccine, the three novel vaccines elicited higher VN levels and reduced injection site inflammation.
Sigma-Aldrich
Dimethyldioctadecylammonium bromide, ≥98% (TLC)
Pricing and availability is not currently available.