Merck
  • Duganella phyllosphaerae sp. nov., isolated from the leaf surface of Trifolium repens and proposal to reclassify Duganella violaceinigra into a novel genus as Pseudoduganella violceinigra gen. nov., comb. nov.
Duganella phyllosphaerae sp. nov., isolated from the leaf surface of Trifolium repens and proposal to reclassify Duganella violaceinigra into a novel genus as Pseudoduganella violceinigra gen. nov., comb. nov.
Systematic and applied microbiology (2011-12-16)
Peter Kämpfer, Stefanie Wellner, Kerstin Lohse, Karin Martin, Nicole Lodders
ABSTRACT
A bright yellow pigmented bacterium was isolated from the leaf surface of Trifolium repens in Germany. Comparative analysis of 16S rRNA gene sequences showed that this bacterium is most closely related to Duganella zoogloeoides IAM 12670(T), with a similarity of 99.3%, but revealed only a moderate similarity (96.8%) to the second Duganella species, Duganella violaceinigra YIM 31327(T). Strain T54(T) is clearly different from D. zoogloeoides IAM 12670(T) in that DNA-DNA hybridization revealed a similarity value of 46% (reciprocal 42%). Ubiquinone (Q-8) was the respiratory quinone and the predominant polar lipids consisted of phosphatidylglycerol, phosphatidylethanolamine, three unknown phospholipids and one aminolipid. Strain T54(T) can be distinguished from D. zoogloeoides by the carbon substrate utilization tests of d-trehalose, cis-aconitate, trans-aconitate, glutarate and dl-3-hydroxybutyrate, and 4-hydroxybenzoate in addition to a different polar lipid profile. The name Duganella phyllosphaerae sp. nov. is proposed for this novel species, with the type strain T54(T) (=LMG 25994 = CCM 7824(T)) [corrected]. In addition, it is proposed to reclassify D. violaceinigra into a novel genus Pseudoduganella gen. nov. as the novel species Pseudoduganella violaceinigra comb. nov. because of the low 16S rRNA gene sequence similarities to the other Duganella species (<97%) and striking differences in chemotaxonomic (lipid profiles and fatty acid patterns) and other phenotypic features, including the colony pigmentation.
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 10 purifications
Pricing and availability is not currently available.