Merck
  • Presence and expression of human papillomavirus sequences in human cervical carcinoma cell lines.
Presence and expression of human papillomavirus sequences in human cervical carcinoma cell lines.
The American journal of pathology (1985-06-01)
C Yee, I Krishnan-Hewlett, C C Baker, R Schlegel, P M Howley
ABSTRACT
A series of human carcinoma cell lines was examined for human papillomavirus (HPV) DNA sequences with the use of HPV-6, HPV-11, HPV-16, and HPV-18 DNA probes. Six of eight cell lines derived from human cervical carcinomas were shown to contain integrated HPV DNA sequences. In five of these six lines, HPV-specific polyadenylated RNA species could also be identified. The expression of HPV sequences was detected in three lines with a HPV-18 DNA probe and in two lines with a HPV-16 DNA probe. Of the two lines which contained HPV-16 specific RNA, one contained HPV DNA sequences which hybridized only to an HPV-16 probe, and the other contained HPV DNA sequences which hybridized to both HPV-16 and HPV-18 DNA probes. Six cell lines established from human squamous-cell carcinomas of the bladder, pharynx, lung, esophagus, and vulva were negative for HPV-6, HPV-11, HPV-16, and HPV-18 DNA sequences under stringent hybridization conditions.
Product Description
HeLa229 cell line
Ca Ski, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87020501, human cervix (small bowel metastasis), Epithelial