Merck
  • Mutations in the gene encoding 3-hydroxyisobutyryl-CoA hydrolase results in progressive infantile neurodegeneration.
Mutations in the gene encoding 3-hydroxyisobutyryl-CoA hydrolase results in progressive infantile neurodegeneration.
American journal of human genetics (2006-12-13)
Ference J Loupatty, Peter T Clayton, Jos P N Ruiter, Rob Ofman, Lodewijk Ijlst, Garry K Brown, David R Thorburn, Robert A Harris, Marinus Duran, Carlos Desousa, Steve Krywawych, Simon J R Heales, Ronald J A Wanders
ABSTRACT
Only a single patient with 3-hydroxyisobutyryl-CoA hydrolase deficiency has been described in the literature, and the molecular basis of this inborn error of valine catabolism has remained unknown until now. Here, we present a second patient with 3-hydroxyisobutyryl-CoA hydrolase deficiency, who was identified through blood spot acylcarnitine analysis showing persistently increased levels of hydroxy-C(4)-carnitine. Both patients manifested hypotonia, poor feeding, motor delay, and subsequent neurological regression in infancy. Additional features in the newly identified patient included episodes of ketoacidosis and Leigh-like changes in the basal ganglia on a magnetic resonance imaging scan. In cultured skin fibroblasts from both patients, the 3-hydroxyisobutyryl-CoA hydrolase activity was deficient, and virtually no 3-hydroxyisobutyryl-CoA hydrolase protein could be detected by western blotting. Molecular analysis in both patients uncovered mutations in the HIBCH gene, including one missense mutation in a conserved part of the protein and two mutations affecting splicing. A carefully interpreted acylcarnitine profile will allow more patients with 3-hydroxyisobutyryl-CoA hydrolase deficiency to be diagnosed.
Sigma-Aldrich
Sodium (R)-β-hydroxyisobutyrate, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
(±)-Sodium β-hydroxyisobutyrate, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium (S)-β-hydroxyisobutyrate, ≥96.0%
Pricing and availability is not currently available.