Merck
  • Signaling events downstream of mammalian target of rapamycin complex 2 are attenuated in cells and tumors deficient for the tuberous sclerosis complex tumor suppressors.
Signaling events downstream of mammalian target of rapamycin complex 2 are attenuated in cells and tumors deficient for the tuberous sclerosis complex tumor suppressors.
Cancer research (2009-07-16)
Jingxiang Huang, Shulin Wu, Chin-Lee Wu, Brendan D Manning
ABSTRACT
Mutations in the TSC1 and TSC2 tumor suppressor genes give rise to the neoplastic disorders tuberous sclerosis complex (TSC) and lymphangioleiomyomatosis. Their gene products form a complex that is a critical negative regulator of mammalian target of rapamycin (mTOR) complex 1 (mTORC1) and cell growth. We recently found that the TSC1-TSC2 complex promotes the activity of mTOR complex 2 (mTORC2), an upstream activator of Akt, and this occurs independent of its inhibitory effects on mTORC1. Loss of mTORC2 activity in cells lacking the TSC1-TSC2 complex, coupled with mTORC1-mediated feedback mechanisms, leads to strong attenuation of the growth factor-stimulated phosphorylation of Akt on S473. In this study, we show that both phosphatidylinositol 3-kinase-dependent and phosphatidylinositol 3-kinase-independent mTORC2 substrates are affected by loss of the TSC1-TSC2 complex in cell culture models and kidney tumors from both Tsc2(+/-) mice (adenoma) and TSC patients (angiomyolipoma). These mTORC2 targets are all members of the AGC kinase family and include Akt, protein kinase Calpha, and serum and glucocorticoid-induced protein kinase 1. We also show that the TSC1-TSC2 complex can directly stimulate the in vitro kinase activity of mTORC2. The interaction between these two complexes is mediated primarily through regions on TSC2 and a core component of mTORC2 called Rictor. Hence, loss of the TSC tumor suppressors results in elevated mTORC1 signaling and attenuated mTORC2 signaling. These findings suggest that the TSC1-TSC2 complex plays opposing roles in tumor progression, both blocking and promoting specific oncogenic pathways through its effects on mTORC1 inhibition and mTORC2 activation, respectively.
Product Description
Millipore
FLAG® Peptide, lyophilized powder