Merck
  • Crystallization of a nonclassical Kazal-type Carcinoscorpius rotundicauda serine protease inhibitor, CrSPI-1, complexed with subtilisin.
Crystallization of a nonclassical Kazal-type Carcinoscorpius rotundicauda serine protease inhibitor, CrSPI-1, complexed with subtilisin.
Acta crystallographica. Section F, Structural biology and crystallization communications (2009-05-02)
Shenoy Rajesh Tulsidas, Saravanan Thangamani, Bow Ho, J Sivaraman, Jeak Ling Ding
ABSTRACT
Serine proteases play a major role in host-pathogen interactions. The innate immune system is known to respond to invading pathogens in a nonspecific manner. The serine protease cascade is an essential component of the innate immune system of the horseshoe crab. The serine protease inhibitor CrSPI isoform 1 (CrSPI-1), a unique nonclassical Kazal-type inhibitor of molecular weight 9.3 kDa, was identified from the hepatopancreas of the horseshoe crab Carcinoscorpius rotundicauda. It potently inhibits subtilisin and constitutes a powerful innate immune defence against invading microbes. Here, the cloning, expression, purification and cocrystallization of CrSPI-1 with subtilisin are reported. The crystals diffracted to 2.6 A resolution and belonged to space group P2(1), with unit-cell parameters a = 73.8, b = 65.0, c = 111.9 A, beta = 95.4 degrees . The Matthews coefficient (V(M) = 2.64 A(3) Da(-1), corresponding to 53% solvent content) and analysis of the preliminary structure solution indicated the presence of one heterotrimer (1:2 ratio of CrSPI-1:subtilisin) and one free subtilisin molecule in the asymmetric unit.
Sigma-Aldrich
Enterokinase human, recombinant, expressed in CHO cells, ≥90% (SDS-PAGE), ≥90% (HPLC), suitable for cell culture
Pricing and availability is not currently available.