Merck
  • Mammalian oocytes store proteins for the early embryo on cytoplasmic lattices.
Mammalian oocytes store proteins for the early embryo on cytoplasmic lattices.
Cell (2023-11-06)
Ida M A Jentoft, Felix J B Bäuerlein, Luisa M Welp, Benjamin H Cooper, Arsen Petrovic, Chun So, Sarah Mae Penir, Antonio Z Politi, Yehor Horokhovskyi, Iina Takala, Heike Eckel, Rüdiger Moltrecht, Peter Lénárt, Tommaso Cavazza, Juliane Liepe, Nils Brose, Henning Urlaub, Rubén Fernández-Busnadiego, Melina Schuh
ABSTRACT
Mammalian oocytes are filled with poorly understood structures called cytoplasmic lattices. First discovered in the 1960s and speculated to correspond to mammalian yolk, ribosomal arrays, or intermediate filaments, their function has remained enigmatic to date. Here, we show that cytoplasmic lattices are sites where oocytes store essential proteins for early embryonic development. Using super-resolution light microscopy and cryoelectron tomography, we show that cytoplasmic lattices are composed of filaments with a high surface area, which contain PADI6 and subcortical maternal complex proteins. The lattices associate with many proteins critical for embryonic development, including proteins that control epigenetic reprogramming of the preimplantation embryo. Loss of cytoplasmic lattices by knocking out PADI6 or the subcortical maternal complex prevents the accumulation of these proteins and results in early embryonic arrest. Our work suggests that cytoplasmic lattices enrich maternally provided proteins to prevent their premature degradation and cellular activity, thereby enabling early mammalian development.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-TLE6 antibody produced in mouse, clone 2E4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-DPPA5 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution