Merck
Methods for Monitoring ER-Phagy.
Methods in molecular biology (Clifton, N.J.) (2024-08-08)
Marisa Di Monaco, Marian Peteri, Jin Rui Liang
ABSTRACT
The endoplasmic reticulum (ER) serves as a central hub for protein synthesis, folding, and lipid biosynthesis in eukaryotic cells. Maintaining ER homeostasis is essential for optimal cellular function, and one mechanism that has garnered attention is endoplasmic reticulum-specific autophagy, or ER-phagy. ER-phagy selectively removes specific ER portions, playing a pivotal role in cellular health and adaptation to environmental stressors. ER-phagy can be induced by diverse cellular conditions such as amino acid starvation, disruption of ER quality control mechanisms, and accumulation of misfolded ER protein, highlighting cellular adaptability and the significance of ER-phagy in stress responses. Clinically relevant mutations in ER-phagy receptors are implicated in various diseases, underlining the fundamental importance of ER-phagy in ER homeostasis. Here, we provide comprehensive protocols and general considerations while investigating ER-phagy using three fundamental techniques-Western blotting, immunofluorescence, and flow cytometry-commonly used in ER-phagy detection and quantitation.
Sigma-Aldrich
Anti-TEX264 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution