Merck
The Azurin Coding Gene: Origin and Phylogenetic Distribution.
Microorganisms (2022-01-22)
Leandro Gammuto, Carolina Chiellini, Marta Iozzo, Renato Fani, Giulio Petroni
ABSTRACT
Azurin is a bacterial-derived cupredoxin, which is mainly involved in electron transport reactions. Interest in azurin protein has risen in recent years due to its anticancer activity and its possible applications in anticancer therapies. Nevertheless, the attention of the scientific community only focused on the azurin protein found in Pseudomonas aeruginosa (Proteobacteria, Gammaproteobacteria). In this work, we performed the first comprehensive screening of all the bacterial genomes available in online repositories to assess azurin distribution in the three domains of life. The Azurin coding gene was not detected in the domains Archaea and Eucarya, whereas it was detected in phyla other than Proteobacteria, such as Bacteroidetes, Verrucomicrobia and Chloroflexi, and a phylogenetic analysis of the retrieved sequences was performed. Observed patchy distribution and phylogenetic data suggest that once it appeared in the bacterial domain, the azurin coding gene was lost in several bacterial phyla and/or anciently horizontally transferred between different phyla, even though a vertical inheritance appeared to be the major force driving the transmission of this gene. Interestingly, a shared conserved domain has been found among azurin members of all the investigated phyla. This domain is already known in P. aeruginosa as p28 domain and its importance for azurin anticancer activity has been widely explored. These findings may open a new and intriguing perspective in deciphering the azurin anticancer mechanisms and to develop new tools for treating cancer diseases.