Merck
  • Alcohol-sourced acetate impairs T cell function by promoting cortactin acetylation.
Alcohol-sourced acetate impairs T cell function by promoting cortactin acetylation.
iScience (2023-07-24)
Vugar Azizov, Michel Hübner, Michael Frech, Jörg Hofmann, Marketa Kubankova, Dennis Lapuente, Matthias Tenbusch, Jochen Guck, Georg Schett, Mario M Zaiss
ABSTRACT
Alcohol is among the most widely consumed dietary substances. Excessive alcohol consumption damages the liver, heart, and brain. Alcohol also has strong immunoregulatory properties. Here, we report how alcohol impairs T cell function via acetylation of cortactin, a protein that binds filamentous actin and facilitates branching. Upon alcohol consumption, acetate, the metabolite of alcohol, accumulates in lymphoid organs. T cells exposed to acetate, exhibit increased acetylation of cortactin. Acetylation of cortactin inhibits filamentous actin binding and hence reduces T cell migration, immune synapse formation and activation. While mutated, acetylation-resistant cortactin rescues the acetate-induced inhibition of T cell migration, primary mouse cortactin knockout T cells exhibited impaired migration. Acetate-induced cytoskeletal changes effectively inhibited activation, proliferation, and immune synapse formation in T cells in vitro and in vivo in an influenza infection model in mice. Together these findings reveal cortactin as a possible target for mitigation of T cell driven autoimmune diseases.
Sigma-Aldrich
Anti-acetyl Cortactin Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.