Merck
  • Multimodal imaging for validation and optimization of ion channel-based chemogenetics in nonhuman primates.
Multimodal imaging for validation and optimization of ion channel-based chemogenetics in nonhuman primates.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2023-08-25)
Yuki Hori, Yuji Nagai, Yukiko Hori, Kei Oyama, Koki Mimura, Toshiyuki Hirabayashi, Ken-Ichi Inoue, Masayuki Fujinaga, Ming-Rong Zhang, Masahiko Takada, Makoto Higuchi, Takafumi Minamimoto
ABSTRACT
Chemogenetic tools provide an opportunity to manipulate neuronal activity and behavior selectively and repeatedly in nonhuman primates (NHPs) with minimal invasiveness. Designer Receptors Exclusively Activated by Designer Drugs are one example that is based on mutated muscarinic acetylcholine receptors. Another channel-based chemogenetic system available for neuronal modulation in NHPs uses Pharmacologically Selective Actuator Modules (PSAMs), which are selectively activated by Pharmacologically Selective Effector Molecules (PSEMs). To facilitate the use of the PSAM/PSEM system, the selection and dosage of PSEMs should be validated and optimized for NHPs. To this end, we used a multimodal imaging approach. We virally expressed excitatory PSAM (PSAM4-5HT3) in the striatum and the primary motor cortex of two male macaque monkeys, and visualized its location through positron emission tomography (PET) with the reporter ligand [18F]ASEM. Chemogenetic excitability of neurons triggered by two PSEMs (uPSEM817 and uPSEM792) was evaluated using [18F]fluorodeoxyglucose-PET imaging, with uPSEM817 being more efficient than uPSEM792. Pharmacological magnetic resonance imaging showed that increased brain activity in the PSAM4-expressing region began approximately 13 min after uPSEM817 administration and continued for at least 60 min. Our multimodal imaging data provide valuable information regarding the manipulation of neuronal activity using the PSAM/PSEM system in NHPs, facilitating future applications.Significance statementLike other chemogenetic tools, the ion channel-based system called Pharmacologically Selective Actuator Module/Pharmacologically Selective Effector Molecule (PSAM/PSEM) allows remote manipulation of neuronal activity and behavior in living animals. Nevertheless, its application in non-human primates (NHPs) is still limited. Here, we used multi-tracer positron emission tomography (PET) imaging and pharmacological magnetic resonance imaging (MRI) to visualize an excitatory chemogenetic ion channel (PSAM4-5HT3) and validate its chemometric function in macaque monkeys. Our results provide the optimal agonist, dose, and timing for chemogenetic neuronal manipulation, facilitating the use of the PSAM/PSEM system and expanding the flexibility and reliability of circuit manipulation in NHPs in a variety of situations.
Sigma-Aldrich
Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4, clone 27-4, from rabbit
Sigma-Aldrich
Anti-DYKDDDDK (FLAG® epitope tag) Antibody, clone 2EL-1B11, ascites fluid, clone 2EL-1B11, Chemicon®