Merck
  • Measuring CTL Lytic Granule Secretion and Target Cell Membrane Repair by Fluorescent Lipophilic Dye Uptake at the Lytic Synapse.
Measuring CTL Lytic Granule Secretion and Target Cell Membrane Repair by Fluorescent Lipophilic Dye Uptake at the Lytic Synapse.
Methods in molecular biology (Clifton, N.J.) (2023-04-28)
Sabina Müller, Liza Filali, Marie-Pierre Puissegur, Salvatore Valitutti
ABSTRACT
CD8+ cytotoxic T lymphocytes (CTL) play a key role in anti-tumor immune response. They are therefore at the heart of current immunotherapy protocols against cancer. Despite current strategies to potentiate CTL responses, cancer cells can resist CTL attack, thus limiting the efficacy of immunotherapies. To optimize immunotherapy, it is urgent to develop rapid assays allowing to assess CTL-cancer cell confrontation at the lytic synapse.In this chapter, we describe a flow cytometry-based method to simultaneously assess the extent of CTL activation and of tumor cell reparative membrane turnover in CTL/target cell conjugates. Such a method can be performed using a limited number of cells. It can therefore be employed in clinical settings when only a few patient-derived cells might be available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Pricing and availability is not currently available.