Merck
  • Giant worm-shaped ESCRT scaffolds surround actin-independent integrin clusters.
Giant worm-shaped ESCRT scaffolds surround actin-independent integrin clusters.
The Journal of cell biology (2023-05-18)
Femmy C Stempels, Muwei Jiang, Harry M Warner, Magda-Lena Moser, Maaike H Janssens, Sjors Maassen, Iris H Nelen, Rinse de Boer, William F Jiemy, David Knight, Julian Selley, Ronan O'Cualain, Maksim V Baranov, Thomas C Q Burgers, Roberto Sansevrino, Dragomir Milovanovic, Peter Heeringa, Matthew C Jones, Rifka Vlijm, Martin Ter Beest, Geert van den Bogaart
ABSTRACT
Endosomal Sorting Complex Required for Transport (ESCRT) proteins can be transiently recruited to the plasma membrane for membrane repair and formation of extracellular vesicles. Here, we discovered micrometer-sized worm-shaped ESCRT structures that stably persist for multiple hours at the plasma membrane of macrophages, dendritic cells, and fibroblasts. These structures surround clusters of integrins and known cargoes of extracellular vesicles. The ESCRT structures are tightly connected to the cellular support and are left behind by the cells together with surrounding patches of membrane. The phospholipid composition is altered at the position of the ESCRT structures, and the actin cytoskeleton is locally degraded, which are hallmarks of membrane damage and extracellular vesicle formation. Disruption of actin polymerization increased the formation of the ESCRT structures and cell adhesion. The ESCRT structures were also present at plasma membrane contact sites with membrane-disrupting silica crystals. We propose that the ESCRT proteins are recruited to adhesion-induced membrane tears to induce extracellular shedding of the damaged membrane.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid