Merck
  • NFATc1 induction by an intronic enhancer restricts NKT γδ cell formation.
NFATc1 induction by an intronic enhancer restricts NKT γδ cell formation.
iScience (2023-03-18)
Sabrina Giampaolo, Cristina M Chiarolla, Konrad Knöpper, Martin Vaeth, Matthias Klein, Azeem Muhammad, Tobias Bopp, Friederike Berberich-Siebelt, Amiya K Patra, Edgar Serfling, Stefan Klein-Hessling
ABSTRACT
In thymus, the ablation of T cell receptor (TCR)-activated transcription factor NFATc1 or its inducible isoforms during the double-negative (DN) stages of thymocyte development leads to a marked increase in γδ thymocytes whereas the development of αβ thymocytes remains mostly unaffected. These γδ thymocytes are characterized by the upregulation of the promyelocytic leukemia zinc-finger factor (PLZF), the "master regulator" of natural killer T (NKT) cell development, and the acquisition of an NKT γδ cell phenotype with higher cell survival rates. The suppressive function of NFATc1 in NKT γδ cell formation critically depends on the remote enhancer E2, which is essential for the inducible expression of NFATc1 directed by its distal promoter P1. Thus, the enhancer deciphers a strong γδ TCR signal into the expression of inducible NFATc1 isoforms resulting in high levels of NFATc1 protein that are essential to control the numbers of NKT γδ cells.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)