Merck
  • Identification of the Highly Active, Species Cross-Reactive Complex I Inhibitor BAY-179.
Identification of the Highly Active, Species Cross-Reactive Complex I Inhibitor BAY-179.
ACS medicinal chemistry letters (2022-03-19)
Jeffrey Mowat, Alexander H M Ehrmann, Sven Christian, Carolyn Sperl, Stephan Menz, Judith Günther, Roman C Hillig, Marcus Bauser, Wolfgang Schwede
ABSTRACT
Mitochondria are key regulators of energy supply and cell death. Generation of ATP within mitochondria occurs through oxidative phosphorylation (OXPHOS), a process which utilizes the four complexes (complex I-IV) of the electron transport chain and ATP synthase. Certain oncogenic mutations (e.g., LKB1 or mIDH) can further enhance the reliance of cancer cells on OXPHOS for their energetic requirements, rendering cells sensitive to complex I inhibition and highlighting the potential value of complex I as a therapeutic target. Herein, we describe the discovery of a potent, selective, and species cross-reactive complex I inhibitor. A high-throughput screen of the Bayer compound library followed by hit triaging and initial hit-to-lead activities led to a lead structure which was further optimized in a comprehensive lead optimization campaign. Focusing on balancing potency and metabolic stability, this program resulted in the identification of BAY-179, an excellent in vivo suitable tool with which to probe the biological relevance of complex I inhibition in cancer indications.
Sigma-Aldrich
Complex 1 probe BAY-179, ≥98% (HPLC)
Pricing and availability is not currently available.