Merck
  • CHK2 Promotes Metabolic Stress-Induced Autophagy through ULK1 Phosphorylation.
CHK2 Promotes Metabolic Stress-Induced Autophagy through ULK1 Phosphorylation.
Antioxidants (Basel, Switzerland) (2022-06-25)
Ran Guo, Shan-Shan Wang, Xiao-You Jiang, Ye Zhang, Yang Guo, Hong-Yan Cui, Qi-Qiang Guo, Liu Cao, Xiao-Chen Xie
ABSTRACT
Reactive oxygen species (ROS) act as a signaling intermediate to promote cellular adaptation to maintain homeostasis by regulating autophagy during pathophysiological stress. However, the mechanism by which ROS promotes autophagy is still largely unknown. Here, we show that the ATM/CHK2/ULK1 axis initiates autophagy to maintain cellular homeostasis by sensing ROS signaling under metabolic stress. We report that ULK1 is a physiological substrate of CHK2, and that the binding of CHK2 to ULK1 depends on the ROS signal and the phosphorylation of threonine 68 of CHK2 under metabolic stress. Further, CHK2 phosphorylates ULK1 on serine 556, and this phosphorylation is dependent on the ATM/CHK2 signaling pathway. CHK2-mediated phosphorylation of ULK1 promotes autophagic flux and inhibits apoptosis induced by metabolic stress. Taken together, these results demonstrate that the ATM/CHK2/ULK1 axis initiates an autophagic adaptive response by sensing ROS, and it protects cells from metabolic stress-induced cellular damage.
Sigma-Aldrich
KU-55933, ≥98% (HPLC)
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Chk2 Inhibitor II hydrate, ≥98% (HPLC)