Merck
  • Metabolic remodeling in tumor-associated macrophages contributing to antitumor activity of cryptotanshinone by regulating TRAF6-ASK1 axis.
Metabolic remodeling in tumor-associated macrophages contributing to antitumor activity of cryptotanshinone by regulating TRAF6-ASK1 axis.
Molecular therapy oncolytics (2022-07-22)
Jia-Hau Yen, Wei-Chieh Huang, Shu-Ching Lin, Yi-Wen Huang, Wan-Ting Chio, Gregory J Tsay, Mien-Chie Hung, Sheng-Teng Huang
ABSTRACT
Dampening tumor growth by converting tumor-associated macrophages (TAMs) from M2/repair-types to M1/kill-types is of high interest. Here, we show that cryptotanshinone (CPT) can function as an antitumor immune modulator that switches TAMs from an M2 to an M1 phenotype, leading to tumor regression. An orthotopic triple-negative breast cancer (TNBC) implantation model was used to determine the role and mechanism of CPT in suppressing M1-to-M2 repolarization of TAMs. Co-culturing TNBC cells with CPT-treated macrophages reduced TNBC proliferation and motility, while in TNBC orthotopic mouse models, CPT treatment inhibited breast tumor formation. Moreover, we identified that CPT inhibits mitochondrial oxidative phosphorylation and mitochondrial fusion via autophagy and transcriptional activation of the apoptosis signal-regulating kinase 1 (ASK1) pathway. Suppression of ASK1 downregulates autophagy and abolishes CPT-induced effects upon TAMs. In addition, CPT inhibits M2 macrophage differentiation and causes TRAF6 auto-ubiquitination-dependent activation of the ASK1, leading to M1 polarization. On the contrary, in M1 macrophage, CPT increases interaction of ASK1 and TRAF6 which induces ASK1 ubiquitination and degradation. Intriguingly, CPT plays opposite roles in the M1 and M2 phenotype. Our findings help to illuminate a previously unrecognized antitumor mechanism of CPT and suggest that this natural compound offers a macrophage-based approach for cancer immunotherapy.
Sigma-Aldrich
Cryptotanshinone, ≥98% (HPLC)
Pricing and availability is not currently available.