Merck
  • Orexin-B-like immunoreactivity in pituitary αMSH-producing cells and median eminence GnRH-containing fibres of the flat-tailed house gecko.
Orexin-B-like immunoreactivity in pituitary αMSH-producing cells and median eminence GnRH-containing fibres of the flat-tailed house gecko.
Anatomia, histologia, embryologia (2019-06-27)
Hirohumi Suzuki, Toshiharu Yamamoto
ABSTRACT
We examined the distribution of the orexin-like peptides in the pituitary and median eminence of the flat-tailed house gecko (Hemidactylus platyurus) using immunohistochemistry. Orexin-B-like, but not orexin-A-like, immunoreactivity was detected in the pituitary, specifically in the pars intermedia, and these cells corresponded to alpha-melanocyte-stimulating hormone (αMSH)-producing cells. Orexin-B and αMSH secreted from pars intermedia may modulate secretion of adenohypophyseal cells in the pars distalis. In the median eminence, orexin-B-immunoreactive puncta and fibres were observed, and these structures corresponded to gonadotropin-releasing hormone (GnRH)-immunoreactive puncta and fibres. Orexin-B secreted from GnRH-containing neurons in the hypothalamus may affect thyrotropin-releasing hormone-containing neurons resulting in modulation of αMSH secretion of melanotrophs in the pars intermedia.
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Melanocyte Stimulating Hormone α Antibody, serum, Chemicon®