Merck
  • Localization of muscarinic receptors M1R, M2R and M3R in the human colon.
Localization of muscarinic receptors M1R, M2R and M3R in the human colon.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2010-02-12)
A M Harrington, C J Peck, L Liu, E Burcher, J M Hutson, B R Southwell
ABSTRACT
Muscarinic acetylcholine receptors (MR) are involved in multiple intestinal reflexes. The cellular localization of subtypes of MRs within enteric circuits mediating muscle and mucosal reflexes remains to be demonstrated. This study aimed to localize the three functionally significant subtypes of MRs in human colon. Reverse transcriptase-PCR was used to determine expression levels of muscarinic receptor subtype (MRs) M1Rs, M2Rs and M3Rs in human colon. Indirect immunofluorescence and confocal microscopy was used to localize MRs in cryostat-cut sections of human colon. Sections were double labeled for multiple cellular and neurochemical markers. Western blotting was used to confirm specificity of the muscarinic antisera used. All three MR subtypes were expressed in human colon. Immunoreactivity (IR) for M2Rs and M3Rs was most abundant in circular and longitudinal muscle. M1R-IR was most abundant on myenteric and submucosal nerve cells, both cholinergic and nitrergic. M3R-IR was also present on populations on myenteric nerve cell bodies. Immunoreactivity for all three receptors was present on nerve fibers in the circular muscle. In the human colon, subtypes of MRs were present on multiple cell types within the enteric circuits underlying motility, secretory and vasoactive reflexes. The cellular distribution for MRs found in this study agrees with data from functional studies, providing insight into the role MRs have in mediating enteric cholinergic neurotransmission.
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m2 Antibody, clone M2-2-B3, clone M2-2-B3, Chemicon®, from rat
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat