Merck
  • A tick C1q protein alters infectivity of the Lyme disease agent by modulating interferon γ.
A tick C1q protein alters infectivity of the Lyme disease agent by modulating interferon γ.
Cell reports (2022-11-24)
Xiaotian Tang, Gunjan Arora, Jaqueline Matias, Thomas Hart, Yingjun Cui, Erol Fikrig
ABSTRACT
In North America, the Lyme disease agent, Borrelia burgdorferi, is commonly transmitted by the black-legged tick, Ixodes scapularis. Tick saliva facilitates blood feeding and enhances pathogen survival and transmission. Here, we demonstrate that I. scapularis complement C1q-like protein 3 (IsC1ql3), a tick salivary protein, directly interacts with B. burgdorferi and is important during the initial stage of spirochetal infection of mice. Mice fed upon by B. burgdorferi-infected IsC1ql3-silenced ticks, or IsC1ql3-immunized mice fed upon by B. burgdorferi-infected ticks, have a lower spirochete burden during the early phase of infection compared with control animals. Mechanically, IsC1ql3 interacts with the globular C1q receptor present on the surface of CD4+ and CD8+ T cells, resulting in decreased production of interferon γ. IsC1ql3 is a C1q-domain-containing protein identified in arthropod vectors and has an important role in B. burgdorferi infectivity as the spirochete transitions from the tick to vertebrate host.
Sigma-Aldrich
Goat Anti-Human IgG Antibody, HRP conjugate, 1.0 mg/mL, Chemicon®
Pricing and availability is not currently available.