Merck
  • Alectinib induces marked red cell spheroacanthocytosis in a near-ubiquitous fashion and is associated with reduced eosin-5-maleimide binding.
Alectinib induces marked red cell spheroacanthocytosis in a near-ubiquitous fashion and is associated with reduced eosin-5-maleimide binding.
Pathology (2021-02-24)
James A Kuzich, Sarah Heynemann, Niall Geoghegan, Cindy Evelyn, Susan O'Mahoney, Susan Wilson, Janine Campbell, Kelly Rogers, Benjamin Solomon, David Westerman, Sant-Rayn Pasricha
ABSTRACT
We reviewed haematological investigations for 43 patients treated at a single centre with alectinib, an inhibitor of anaplastic lymphoma kinase (ALK) which is considered standard first-line treatment for patients with ALK-rearranged advanced non-small cell lung cancer. Ninety-five percent of patients developed marked acanthocytosis, echinocytosis and/or spheroacanthocytosis, not observable with prior treatment with other ALK-inhibitors. Anaemia developed in 73% of patients (38% <100 g/L, 8% <80 g/L), though definite new haemolysis was present in only 11%. Eosin-5-maleimide binding was reduced in all assessed patients, and increased membrane cholesterol was identified in one patient assessed with lattice light sheet microscopy. We have identified a previously undescribed phenomenon whereby alectinib induces red cell membrane abnormalities in nearly all patients through an unclear, but likely ALK-independent, mechanism, resulting in mild anaemia without universal haemolysis.
Sigma-Aldrich
5-Maleimido-eosin, for fluorescence, ≥93% (HPLC)