Merck
  • The HUVEC/Matrigel assay: an in vivo assay of human angiogenesis suitable for drug validation.
The HUVEC/Matrigel assay: an in vivo assay of human angiogenesis suitable for drug validation.
Methods in molecular biology (Clifton, N.J.) (2006-12-19)
Dag K Skovseth, Axel M Küchler, Guttorm Haraldsen
ABSTRACT
The future ability to manipulate the growth of new blood vessels (angiogenesis) holds great promise for treating ischemic disease and cancer. Several models of human in vivo angiogenesis have been described, but they seem to depend on transgenic support and have not been validated in a therapeutic context. Here, we describe an in vivo assay that mimics human angiogenesis in which native human umbilical vein-derived endothelial cells are suspended in a liquid laminin/collagen gel (Matrigel), injected into immunodeficient mice, and develop into mature, functional vessels that vascularize the Matrigel plug in the course of 30 d. Moreover, we demonstrate how to target this process therapeutically by sustained delivery of the angiogenesis inhibitor endostatin from subcutaneously implanted microosmotic pumps.
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
Pricing and availability is not currently available.