Merck
  • Increased superoxide in vivo accelerates age-associated muscle atrophy through mitochondrial dysfunction and neuromuscular junction degeneration.
Increased superoxide in vivo accelerates age-associated muscle atrophy through mitochondrial dysfunction and neuromuscular junction degeneration.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2009-12-31)
Youngmok C Jang, Michael S Lustgarten, Yuhong Liu, Florian L Muller, Arunabh Bhattacharya, Hanyu Liang, Adam B Salmon, Susan V Brooks, Lisa Larkin, Christopher R Hayworth, Arlan Richardson, Holly Van Remmen
ABSTRACT
Oxidative stress has been implicated in the etiology of age-related muscle loss (sarcopenia). However, the underlying mechanisms by which oxidative stress contributes to sarcopenia have not been thoroughly investigated. To directly examine the role of chronic oxidative stress in vivo, we used a mouse model that lacks the antioxidant enzyme CuZnSOD (Sod1). Sod1(-/-) mice are characterized by high levels of oxidative damage and an acceleration of sarcopenia. In the present study, we demonstrate that muscle atrophy in Sod1(-/-) mice is accompanied by a progressive decline in mitochondrial bioenergetic function and an elevation of mitochondrial generation of reactive oxygen species. In addition, Sod1(-/-) muscle exhibits a more rapid induction of mitochondrial-mediated apoptosis and loss of myonuclei. Furthermore, aged Sod1(-/-) mice show a striking increase in muscle mitochondrial content near the neuromuscular junctions (NMJs). Despite the increase in content, the function of mitochondria is significantly impaired, with increased denervated NMJs and fragmentation of acetylcholine receptors. As a consequence, contractile force in aged Sod1(-/-) muscles is greatly diminished. Collectively, we show that Sod1(-/-) mice display characteristics of normal aging muscle in an accelerated manner and propose that the superoxide-induced NMJ degeneration and mitochondrial dysfunction are potential mechanisms of sarcopenia.
Sigma-Aldrich
Anti-Myosin-2 (MYH2) Antibody, clone SC-71, clone SC-71, from mouse
Pricing and availability is not currently available.