Merck
  • The Novel Protein ADAMTS16 Promotes Gastric Carcinogenesis by Targeting IFI27 through the NF-κb Signaling Pathway.
The Novel Protein ADAMTS16 Promotes Gastric Carcinogenesis by Targeting IFI27 through the NF-κb Signaling Pathway.
International journal of molecular sciences (2022-10-15)
Tuoyang Li, Junyi Zhou, Yingming Jiang, Yandong Zhao, Jintuan Huang, Weiyao Li, Zhenze Huang, Zijian Chen, Xiaocheng Tang, Hao Chen, Zuli Yang
ABSTRACT
A disintegrin and metalloproteinase with thrombospondin motifs 16 (ADAMTS16) has been reported to be involved in the pathogenesis of solid cancers. However, its role in gastric cancer (GC) is unclear. In this study, the role of ADAMTS16 in gastric cancer was investigated. The effects of ADAMTS16 on cell migration, invasion, and proliferation were investigated by functional experiments in vivo and in vitro. Downstream signal pathways of ADAMTS16 were confirmed by using bioinformatics analysis, co-immunoprecipitation, and immunofluorescence. Meanwhile, bioinformatics analysis, qRT-PCR, western blot, and dual-luciferase reporter gene analysis assays were used to identify ADAMTS16 targets. The expression of ADAMTS16 in GC was analyzed in public datasets. The expression of ADAMTS16 and its correlations with the clinical characteristics of GC were investigated by immunohistochemistry. Ectopic ADAMTS16 expression significantly promoted tumor cell migration, invasion, and growth. Bioinformatics analysis and western blot showed that ADAMTS16 upregulated the IFI27 protein through the NF-κb pathway, which was confirmed by immunofluorescence and western blot. Dual-luciferase reporter gene analysis identified a binding site between P65 and IFI27 that may be directly involved in the transcriptional regulation of IFI27. IFI27 knockdown reversed the promoting effect of ADAMTS16 on cell invasion, migration, and proliferation indicating that ADAMTS16 acts on GC cells by targeting the NF-κb/IFI27 axis. ADAMTS16 was associated with poor prognosis in clinical characteristics. ADAMTS16 promotes cell migration, invasion, and proliferation by targeting IFI27 through the NF-κB pathway and is a potential progressive and survival biomarker of GC.
Sigma-Aldrich
Anti-IFI27 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)