Merck
β(2) -adrenoceptor agonists: current and future direction.
British journal of pharmacology (2011-01-15)
Mario Cazzola, Luigino Calzetta, Maria Gabriella Matera
ABSTRACT
Despite the passionate debate over the use of β(2) -adrenoceptor agonists in the treatment of airway disorders, these agents are still central in the symptomatic management of asthma and COPD. A variety of β(2) -adrenoceptor agonists with long half-lives, also called ultra long-acting β(2) -adrenoceptor agonists (ultra-LABAs; indacaterol, olodaterol, vilanterol, carmoterol, LAS100977 and PF-610355) are currently under development with the hopes of achieving once-daily dosing. It is likely that the once-daily dosing of a bronchodilator would be a significant convenience and probably a compliance-enhancing advantage, leading to improved overall clinical outcomes. As combination therapy with an inhaled corticosteroid (ICS) and a LABA is important for treating patients suffering from asthma, and a combination with an inhaled long-acting antimuscarinic agent (LAMA) is important for treating COPD patients whose conditions are not sufficiently controlled by monotherapy with a β(2) -adrenoceptor agonist, some novel once-daily combinations of LABAs and ICSs or LAMAs are under development.
Sigma-Aldrich
Vilanterol trifenatate, ≥95% (HPLC)