Merck
  • Single-cell multiomics analyses of spindle-transferred human embryos suggest a mostly normal embryonic development.
Single-cell multiomics analyses of spindle-transferred human embryos suggest a mostly normal embryonic development.
PLoS biology (2022-08-17)
Shuyue Qi, Wei Wang, Xiaohui Xue, Zhuo Lu, Jia Yan, Yunfei Li, Yu Zhang, Mingming Shu, Chunlan Song, Qihang Wang, Yunhai Chuai, Xinyu Zhai, Shujie Han, Fuchou Tang, Wei Shang
ABSTRACT
Mitochondrial DNA (mtDNA) mutations are often associated with incurable diseases and lead to detectable pathogenic variants in 1 out of 200 babies. Uncoupling of the inheritance of mtDNA and the nuclear genome by spindle transfer (ST) can potentially prevent the transmission of mtDNA mutations from mother to offspring. However, no well-established studies have critically assessed the safety of this technique. Here, using single-cell triple omics sequencing method, we systematically analyzed the genome (copy number variation), DNA methylome, and transcriptome of ST and control blastocysts. The results showed that, compared to that in control embryos, the percentage of aneuploid cells in ST embryos did not significantly change. The epiblast, primitive endoderm, and trophectoderm (TE) of ST blastocysts presented RNA expression profiles that were comparable to those of control blastocysts. However, the DNA demethylation process in TE cells of ST blastocysts was slightly slower than that in the control blastocysts. Collectively, our results suggest that ST seems generally safe for embryonic development, with a relatively minor delay in the DNA demethylation process at the blastocyst stage.
Sigma-Aldrich
Cytochalasin B, Helminthosporium dematioideum, Cell-permeable fungal toxin.
Pricing and availability is not currently available.