- Generation of human parallel chimeric antigen receptor (pCAR) T cells to achieve synergistic T cell co-stimulation.
Generation of human parallel chimeric antigen receptor (pCAR) T cells to achieve synergistic T cell co-stimulation.
Dual co-stimulation may be harnessed using parallel chimeric antigen receptors (pCARs) in which two distinct co-stimulatory units are adjacently localized on the plasma membrane. This protocol summarizes construct design, human T cell isolation, retroviral transduction, tissue culture expansion, and preclinical testing of pCAR T cells, exemplified by receptors that co-target avb6 integrin and ErbB dimers. For complete details on the use and execution of this protocol, please refer to Muliaditan et al. (2021).