Merck
  • The vestigial Quadrant Enhancer is dispensable for pattern formation and development of the Drosophila wing.
The vestigial Quadrant Enhancer is dispensable for pattern formation and development of the Drosophila wing.
microPublication biology (2022-07-06)
Keity J Farfán-Pira, Teresa I Martínez-Cuevas, Rosalio Reyes, Timothy A Evans, Marcos Nahmad
ABSTRACT
In Drosophila , the pattern of the wing selector gene, vestigial ( vg ), is established by at least two enhancers: the Boundary Enhancer, which drives expression along the disc's Dorsal-Ventral boundary; and the Quadrant Enhancer (QE) that patterns the rest of the wing pouch. Using CRISPR/Cas9 editing, we deleted DNA fragments around the reported QE sequence and found that the full Vg pattern is formed. Furthermore, adult wings arising from these gene-edited animals are normal in shape and pattern, but slightly smaller in size, although this reduction is not wing-specific in males. We suggest that other enhancers act redundantly to establish the vg pattern and rescue wing development.
Sigma-Aldrich
PIPES sodium salt, ≥99% (titration)