Merck
  • α-catenin switches between a slip and an asymmetric catch bond with F-actin to cooperatively regulate cell junction fluidity.
α-catenin switches between a slip and an asymmetric catch bond with F-actin to cooperatively regulate cell junction fluidity.
Nature communications (2022-03-05)
C Arbore, M Sergides, L Gardini, G Bianchi, A V Kashchuk, I Pertici, P Bianco, F S Pavone, M Capitanio
ABSTRACT
α-catenin is a crucial protein at cell junctions that provides connection between the actin cytoskeleton and the cell membrane. At adherens junctions (AJs), α-catenin forms heterodimers with β-catenin that are believed to resist force on F-actin. Outside AJs, α-catenin forms homodimers that regulates F-actin organization and directly connect the cell membrane to the actin cytoskeleton, but their mechanosensitive properties are inherently unknown. By using ultra-fast laser tweezers we found that a single α-β-catenin heterodimer does not resist force but instead slips along F-actin in the direction of force. Conversely, the action of 5 to 10 α-β-catenin heterodimers together with force applied toward F-actin pointed end engaged a molecular switch in α-catenin, which unfolded and strongly bound F-actin as a cooperative catch bond. Similarly, an α-catenin homodimer formed an asymmetric catch bond with F-actin triggered by protein unfolding under force. Our data suggest that α-catenin clustering together with intracellular tension engage a fluid-to-solid phase transition at the membrane-cytoskeleton interface.