Merck
  • Patterns of transcription factor programs and immune pathway activation define four major subtypes of SCLC with distinct therapeutic vulnerabilities.
Patterns of transcription factor programs and immune pathway activation define four major subtypes of SCLC with distinct therapeutic vulnerabilities.
Cancer cell (2021-01-23)
Carl M Gay, C Allison Stewart, Elizabeth M Park, Lixia Diao, Sarah M Groves, Simon Heeke, Barzin Y Nabet, Junya Fujimoto, Luisa M Solis, Wei Lu, Yuanxin Xi, Robert J Cardnell, Qi Wang, Giulia Fabbri, Kasey R Cargill, Natalie I Vokes, Kavya Ramkumar, Bingnan Zhang, Carminia M Della Corte, Paul Robson, Stephen G Swisher, Jack A Roth, Bonnie S Glisson, David S Shames, Ignacio I Wistuba, Jing Wang, Vito Quaranta, John Minna, John V Heymach, Lauren Averett Byers
ABSTRACT
Despite molecular and clinical heterogeneity, small cell lung cancer (SCLC) is treated as a single entity with predictably poor results. Using tumor expression data and non-negative matrix factorization, we identify four SCLC subtypes defined largely by differential expression of transcription factors ASCL1, NEUROD1, and POU2F3 or low expression of all three transcription factor signatures accompanied by an Inflamed gene signature (SCLC-A, N, P, and I, respectively). SCLC-I experiences the greatest benefit from the addition of immunotherapy to chemotherapy, while the other subtypes each have distinct vulnerabilities, including to inhibitors of PARP, Aurora kinases, or BCL-2. Cisplatin treatment of SCLC-A patient-derived xenografts induces intratumoral shifts toward SCLC-I, supporting subtype switching as a mechanism of acquired platinum resistance. We propose that matching baseline tumor subtype to therapy, as well as manipulating subtype switching on therapy, may enhance depth and duration of response for SCLC patients.
Sigma-Aldrich
5-Aza-2′-Deoxycytidine, A cytosine analog that acts as a DNA methyltransferase inhibitor.
Sigma-Aldrich
GSK-LSD1, ≥98% (HPLC)