Merck
  • Effect of Long-Term Consumption of Poultry Egg Products on Growth, Body Composition, and Liver Gene Expression in Zebrafish, Danio rerio.
Effect of Long-Term Consumption of Poultry Egg Products on Growth, Body Composition, and Liver Gene Expression in Zebrafish, Danio rerio.
Current developments in nutrition (2022-01-08)
Michael B Williams, Joseph W Palmer, Sophie B Chehade, Alex J Hall, Robert J Barry, Mickie L Powell, Melissa L Harris, Liou Y Sun, Stephen A Watts
ABSTRACT
Poultry eggs are a low-cost, high-protein nutrient package that can be consumed as part of quality diets. However, consumption of poultry egg products is historically contentious, which highlights the importance of investigating impacts of long-term egg consumption on metabolic health. Our study utilized the zebrafish, Danio rerio, a newly defined model of human metabolic health, to understand the metabolic consequence of consuming egg products in lieu of other well-described protein sources. Reference diets were formulated to contain multisource protein with casein and fish protein hydrolysate (CON; control protein sources), the protein sources that have been historically utilized in numerous reference diets. These proteins were then partially replaced with either whole egg (WE; protein and lipid source), egg white (EW; protein source), wheat gluten (WG; cereal protein source), or a high-lipid-content diet containing a multisource protein with casein and fish protein hydrolysate (HFCON; isonitrogenous and isolipidic with the WE diet) in a 34-wk trial (n = 8 tanks, 10 fish per tank). Daily feeding was initiated at the early juvenile life stage and terminated at the late reproductive adult stage. The amino acid composition of control versus egg product diets did not vary substantially, although methionine and lysine were apparently limiting in fish fed WG. At termination, fish fed EW as the protein source had weight gain and body composition similar to those fed the CON diet. Fasting and postprandial blood glucose did not differ between any dietary treatment. Assessment of the liver transcriptome using RNAseq revealed no differential gene expression between zebrafish fed CON or WE diets. Zebrafish fed WG had lower weight gain in males. Long-term consumption of egg products promoted metabolic health equal to that of historically relevant proteins. These data support the value of egg products for maintaining long-term metabolic health in animal diets.
Sigma-Aldrich
Gluten from wheat