Merck
  • Aldolase and actin protect rabbit muscle lactate dehydrogenase from ascorbate inhibition.
Aldolase and actin protect rabbit muscle lactate dehydrogenase from ascorbate inhibition.
Journal of enzyme inhibition and medicinal chemistry (2004-06-19)
Percy J Russell, Anita Williams, Xavier Amador, Reynaldo Vargas
ABSTRACT
Muscle-type LDH (LDH-m4) activity is critical for efficient anaerobic glycolysis. The results here show that rabbit LDH-M4 is inhibited by concentrations of ascorbate normally found in tissues. Aldolase and muscle G-actin were found to protect and to reverse inhibitions of LDH-m4 by ascorbate. G-actins showed some species specificity. Myosin, tropomyosin and troponin from rabbit muscle and muscle proteins from other animal sources had no affect on the inhibitions by ascorbate. The substrate inhibition of LDH-m4 by pyruvate is partially relieved by the presence of aldolase and lowers the Km without affecting the Vm. G-actin under similar conditions has no affect. It is believed that these studies reflect some of the resting properties of glycolytic enzymes that bind and unbind to contractile elements. It is proposed that ascorbate facilitates the storage of glycogen in muscle at rest by inhibiting glycolysis.
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Pyruvate Kinase/Lactic Dehydrogenase enzymes from rabbit muscle, For the Determination of ADP, buffered aqueous glycerol solution
Pricing and availability is not currently available.