Merck
  • Telmisartan Protects Auditory Hair Cells from Gentamicin-Induced Toxicity in vitro.
Telmisartan Protects Auditory Hair Cells from Gentamicin-Induced Toxicity in vitro.
Audiology & neuro-otology (2020-05-06)
Maurizio Cortada, Eric Wei, Neha Jain, Soledad Levano, Daniel Bodmer
ABSTRACT
Telmisartan is an angiotensin II receptor blocker that has pleiotropic effects and protective properties in different cell types. Moreover, telmisartan has also shown partial agonism on the peroxisome proliferator-activated receptor γ (PPAR-γ). Auditory hair cells (HCs) express PPAR-γ, and the protective role of PPAR-γ agonists on HCs has been shown. The objective of this study was to investigate the effects of telmisartan on gentamicin-induced ototoxicity in vitro. Cochlear explants were exposed to gentamicin with or without telmisartan, and/or GW9662, an irreversible PPAR-γ antagonist. Telmisartan protected auditory HCs against gentamicin-induced ototoxicity. GW9662 completely blocked this protective effect, suggesting that it was mediated by PPAR-γ signaling. Exposure to GW9662 or telmisartan alone was not toxic to auditory HCs. We found that telmisartan, via PPAR-γ signaling, protects auditory HCs from gentamicin-induced ototoxicity. Therefore, telmisartan could potentially be used in the future to prevent or treat sensorineural hearing loss.
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Sigma-Aldrich
GW9662, >98% (HPLC)