Merck
  • A novel luminescence-based β-arrestin recruitment assay for unmodified receptors.
A novel luminescence-based β-arrestin recruitment assay for unmodified receptors.
The Journal of biological chemistry (2021-03-09)
Maria Hauge Pedersen, Jennifer Pham, Helena Mancebo, Asuka Inoue, Wesley B Asher, Jonathan A Javitch
ABSTRACT
G protein-coupled receptors (GPCRs) signal through activation of G proteins and subsequent modulation of downstream effectors. More recently, signaling mediated by β-arrestin has also been implicated in important physiological functions. This has led to great interest in the identification of biased ligands that favor either G protein or β-arrestin-signaling pathways. However, nearly all screening techniques for measuring β-arrestin recruitment have required C-terminal receptor modifications that can in principle alter protein interactions and thus signaling. Here, we have developed a novel luminescence-based assay to measure β-arrestin recruitment to the membrane or early endosomes by unmodified receptors. Our strategy uses NanoLuc, an engineered luciferase from Oplophorus gracilirostris (deep-sea shrimp) that is smaller and brighter than other well-established luciferases. Recently, several publications have explored functional NanoLuc split sites for use in complementation assays. We have identified a unique split site within NanoLuc and fused the corresponding N-terminal fragment to either a plasma membrane or early endosome tether and the C-terminal fragment to β-arrestins, which form the basis for the MeNArC and EeNArC assays, respectively. Upon receptor activation, β-arrestin is recruited to the membrane and subsequently internalized in an agonist concentration-dependent manner. This recruitment promotes complementation of the two NanoLuc fragments, thereby reconstituting functional NanoLuc, allowing for quantification of β-arrestin recruitment with a single luminescence signal. Our assay avoids potential artifacts related to C-terminal receptor modification and has promise as a new generic assay for measuring β-arrestin recruitment to diverse GPCR types in heterologous or native cells.
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder