Merck
  • Isolation and characterization of Sprague-Dawley and Wistar Kyoto GFP rat embryonic stem cells.
Isolation and characterization of Sprague-Dawley and Wistar Kyoto GFP rat embryonic stem cells.
Stem cell research (2017-07-06)
Giulia Coppiello, Gloria Abizanda, Natalia Aguado, Elena Iglesias, Olalla Iglesias-Garcia, Antonio Lo Nigro, Felipe Prosper, Xabier L Aranguren
ABSTRACT
We generated two rat embryonic stem cell (ESC) lines: ATCe-SD7.8 from Sprague-Dawley strain and ATCe-WK1 from Wistar Kyoto strain. Cells were marked with enhanced green fluorescent protein (eGFP) by transduction with a lentiviral vector. Cells present a normal karyotype and express pluripotency-associated markers. Pluripotency was tested in vivo with the teratoma formation assay. Cells maintain eGFP expression upon differentiation to the three-germ layers. These cells can be a useful tool for cell therapy studies and chimera generation as they can be easily tracked by eGFP expression.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Pricing and availability is not currently available.