- A signaling pathway governing early thymocyte maturation.
A signaling pathway governing early thymocyte maturation.
Immunology today (1995-02-01)
S J Anderson, R M Perlmutter
PMID7888074
ABSTRACT
Recent evidence supports the view that p56lck, a non-receptor protein tyrosine kinase, serves as the signaling element that 'senses' synthesis of a functional T-cell receptor beta chain, thereby promoting thymocyte maturation. Here, Steven Anderson and Roger Perlmutter review current data, and outline the features of this developmental program.