Merck
  • Polyphyllin I Inhibits Propionibacterium acnes-Induced IL-8 Secretion in HaCaT Cells by Downregulating the CD36/NOX1/ROS/NLRP3/IL-1β Pathway.
Polyphyllin I Inhibits Propionibacterium acnes-Induced IL-8 Secretion in HaCaT Cells by Downregulating the CD36/NOX1/ROS/NLRP3/IL-1β Pathway.
Evidence-based complementary and alternative medicine : eCAM (2021-10-05)
Shuyun Yang, Ying Jiang, Xiuqin Yu, Liping Zhu, Lu Wang, Jingzhu Mao, Miaomiao Wang, Naihui Zhou, Ziliang Yang, Ying Liu, Tingting Zhu
ABSTRACT
Acne vulgaris (AV) is a chronic skin disease involving inflammation of the pilosebaceous units. Propionibacterium acnes (P. acnes) hypercolonization is one pathogenic factor for AV. P. acnes that triggers interleukin-1β (IL-1β) by activating the pyrin domain-containing 3 protein (NLRP3) inflammasome of the NOD-like receptor family in human monocytes. Reactive oxygen species (ROS) acts as a trigger for the production of IL-8 and activates theNLRP3 inflammasome. IL-8 promotes the metastasis and multiplication of different cancerous cells, whereas keratinocyte proliferation and migration contribute to the progression of AV. A steroidal saponin called polyphyllin I (PPI) that is extracted from Paris polyphylla's rhizomes has anti-inflammatory properties. This study investigates the regulatory role of P. acnes in the secretion of IL-8 mediated by the CD36/NADPH oxidase 1 (NOX1)/ROS/NLRP3/IL-1β pathway and the effects of PPI on the CD36/NOX1/ROS/NLRP3/IL-1β/IL-8 pathway and human keratinocyte proliferation and migration. HaCaT cells were cultured and stimulated with 108 CFU/ml of P. acnes for 0, 6, 12, 18, 24, 30, and 36 hours. P. acnes induced IL-8 secretion from HaCaT cells via the CD36/NOX1/ROS/NLRP3/IL-1β pathway. PPI inhibited the CD36/NLRP3/NOX1/ROS/IL-8/IL-1β pathway and HaCaT cell proliferation and migration. PPI alleviates P. acnes-induced inflammatory responses and human keratinocyte proliferation and migration, implying a novel potential therapy for AV.
Sigma-Aldrich
Anti-Ipaf (NLRC4) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-ASC Antibody, Chemicon®, from rabbit