Merck
  • Macrophage polarization-associated lnc-Ma301 interacts with caprin-1 to inhibit hepatocellular carcinoma metastasis through the Akt/Erk1 pathway.
Macrophage polarization-associated lnc-Ma301 interacts with caprin-1 to inhibit hepatocellular carcinoma metastasis through the Akt/Erk1 pathway.
Cancer cell international (2021-08-12)
Hong-Lin Luo, Tao Luo, Jun-Jie Liu, Fei-Xiang Wu, Tao Bai, Chao Ou, Jie Chen, Le-Qun Li, Jian-Hong Zhong
ABSTRACT
Epithelial-mesenchymal transition (EMT) promotes migration, invasion, and metastasis of hepatocellular carcinoma (HCC) cells. The molecular mechanisms behind EMT and metastasis in HCC remain unclear. Microarray analysis was used to identify lncRNAs expression during polarization of U937 macrophages from M2 to M1 phenotype. The expression of the identified lncRNA was compared between clinical samples of HCC tissues or adjacent normal tissues, as well as between HCC and normal liver cell lines. lnc-Ma301 was overexpressed or knocked-down in HCC cell lines, and the effects were assessed in vitro and in vivo. Interactions among lnc-Ma301 and its potential downstream targets caprin-1 were investigated in HCC cell lines. Effects of lnc-Ma301 over- and underexpression on the Akt/Erk1 signaling pathways were examined. Microarray analyses identified lnc-Ma301 as one of the most overexpressed long non-coding RNAs during polarization of U937 macrophages from M2 to M1 phenotype. Lnc-Ma301 showed lower expression in HCC tissues than in adjacent normal tissues, and lower expression was associated with worse prognosis. Activation of lnc-Ma301 inhibited cell proliferation, migration and EMT in HCC cell cultures, and it inhibited lung metastasis of HCC tumors in mice. Mechanistic studies suggested that lnc-Ma301 interacts with caprin-1 to inhibit HCC metastasis and EMT through Akt/Erk1 pathway. Lnc-Ma301 may help regulate onset and metastasis of HCC.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Pricing and availability is not currently available.
Sigma-Aldrich
IFN- γ Protein, Human Recombinant Animal Free, The Interferon gamma (or IFN-γ protein) is an acid-labile interferon produced by CD4 & CD8 T lymphocytes as well as activated NK cells. Manufactured using all non-animal reagents.
Pricing and availability is not currently available.
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Pricing and availability is not currently available.