Merck
  • Discovery and Characterization of R/S-N-3-Cyanophenyl-N'-(6-tert-butoxycarbonylamino-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran-4-yl)urea, a New Histone Deacetylase Class III Inhibitor Exerting Antiproliferative Activity against Cancer Cell Lines.
Discovery and Characterization of R/S-N-3-Cyanophenyl-N'-(6-tert-butoxycarbonylamino-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran-4-yl)urea, a New Histone Deacetylase Class III Inhibitor Exerting Antiproliferative Activity against Cancer Cell Lines.
Journal of medicinal chemistry (2017-05-06)
Michael Schnekenburger, Eric Goffin, Jin-Young Lee, Jun Young Jang, Aloran Mazumder, Seungwon Ji, Bernard Rogister, Nafila Bouider, Florence Lefranc, Walter Miklos, Véronique Mathieu, Pascal de Tullio, Kyu-Won Kim, Mario Dicato, Walter Berger, Byung Woo Han, Robert Kiss, Bernard Pirotte, Marc Diederich
ABSTRACT
A new series of N-aryl-N'-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran-4-yl)ureas bearing an alkoxycarbonylamino group at the 6-position were synthesized and examined as putative anticancer agents targeting sirtuins in glioma cells. On the basis of computational docking combined to in vitro sirtuin 1/2 inhibition assays, we selected compound 18 [R/S-N-3-cyanophenyl-N'-(6-tert-butoxycarbonylamino-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran-4-yl)urea] which displays a potent antiproliferative activity on various glioma cell types, assessed by quantitative videomicroscopy, eventually triggering senescence. The impact on normal glial cells was lower with a selectivity index of >10. Furthermore, human U373 and Hs683 glioblastoma cell lines served to demonstrate the inhibitory activity of 18 against histone deacetylase (HDAC) class III sirtuins 1 and 2 (SIRT1/2) by quantifying acetylation levels of histone and non-histone proteins. The translational potential of 18 was validated by an NCI-60 cell line screen and validation of growth inhibition of drug resistant cancer cell models. Eventually, the anticancer potential of 18 was validated in 3D glioblastoma spheroids and in vivo by zebrafish xenografts. In summary, compound 18 is the first representative of a new class of SIRT inhibitors opening new perspectives in the medicinal chemistry of HDAC inhibitors.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Histone H1 Antibody, clone AE-4, clone AE-4, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H4 Antibody, pan, clone 62-141-13, rabbit monoclonal, clone 62-141-13, Upstate®, from rabbit