Merck
  • Order and disorder-An integrative structure of the full-length human growth hormone receptor.
Order and disorder-An integrative structure of the full-length human growth hormone receptor.
Science advances (2021-07-02)
Noah Kassem, Raul Araya-Secchi, Katrine Bugge, Abigail Barclay, Helena Steinocher, Adree Khondker, Yong Wang, Aneta J Lenard, Jochen Bürck, Cagla Sahin, Anne S Ulrich, Michael Landreh, Martin Cramer Pedersen, Maikel C Rheinstädter, Per Amstrup Pedersen, Kresten Lindorff-Larsen, Lise Arleth, Birthe B Kragelund
ABSTRACT
Because of its small size (70 kilodalton) and large content of structural disorder (>50%), the human growth hormone receptor (hGHR) falls between the cracks of conventional high-resolution structural biology methods. Here, we study the structure of the full-length hGHR in nanodiscs with small-angle x-ray scattering (SAXS) as the foundation. We develop an approach that combines SAXS, x-ray diffraction, and NMR spectroscopy data obtained on individual domains and integrate these through molecular dynamics simulations to interpret SAXS data on the full-length hGHR in nanodiscs. The hGHR domains reorient freely, resulting in a broad structural ensemble, emphasizing the need to take an ensemble view on signaling of relevance to disease states. The structure provides the first experimental model of any full-length cytokine receptor in a lipid membrane and exemplifies how integrating experimental data from several techniques computationally may access structures of membrane proteins with long, disordered regions, a widespread phenomenon in biology.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), peroxidase conjugate, lyophilized powder
Pricing and availability is not currently available.