Merck
  • Brief report: Tempol, a novel antioxidant, inhibits both activated T cell and antigen presenting cell derived cytokines in-vitro from COVID-19 patients.
Brief report: Tempol, a novel antioxidant, inhibits both activated T cell and antigen presenting cell derived cytokines in-vitro from COVID-19 patients.
Clinical immunology (Orlando, Fla.) (2021-08-24)
Kavita Mathi, Yael Rosenberg-Hasson, Holden Maecker, Dennis J Carlo, Ronald B Moss
ABSTRACT
COVID-19 is characterized by a dysregulation of inflammatory cytokines ultimately resulting a cytokine storm that can result in significant morbidity and mortality. We developed an in-vitro assay using activated peripheral blood mononuclear cells (PBMCs) stimulated with lipopolysaccharide (LPS) or CD3 + CD28 to examine secretion of cytokines from antigen presenting cells (APCs) and T cells, respectively, in donor patients with a history of COVID-19 (convalescent) and uninfected negative controls. We hypothesized that a novel antioxidant called Tempol may decrease cytokines from activated peripheral blood cells from both COVID-19 patients and normal donors. Preincubation of immune cells with Tempol resulted in a significant (P < 0.05) decrease in multiple T cell and APC-derived cytokines from both cells of COVID-19 (n = 7) and uninfected donors (n = 7). These preliminary results suggest that Tempol has strong in-vitro anti-cytokine activity and supports additional studies examining the use of Tempol for the treatment of COVID-19.