Merck
TALEN-based gene correction for epidermolysis bullosa.
Molecular therapy : the journal of the American Society of Gene Therapy (2013-04-03)
Mark J Osborn, Colby G Starker, Amber N McElroy, Beau R Webber, Megan J Riddle, Lily Xia, Anthony P DeFeo, Richard Gabriel, Manfred Schmidt, Christof von Kalle, Daniel F Carlson, Morgan L Maeder, J Keith Joung, John E Wagner, Daniel F Voytas, Bruce R Blazar, Jakub Tolar
ABSTRACT
Recessive dystrophic epidermolysis bullosa (RDEB) is characterized by a functional deficit of type VII collagen protein due to gene defects in the type VII collagen gene (COL7A1). Gene augmentation therapies are promising, but run the risk of insertional mutagenesis. To abrogate this risk, we explored the possibility of using engineered transcription activator-like effector nucleases (TALEN) for precise genome editing. We report the ability of TALEN to induce site-specific double-stranded DNA breaks (DSBs) leading to homology-directed repair (HDR) from an exogenous donor template. This process resulted in COL7A1 gene mutation correction in primary fibroblasts that were subsequently reprogrammed into inducible pluripotent stem cells and showed normal protein expression and deposition in a teratoma-based skin model in vivo. Deep sequencing-based genome-wide screening established a safety profile showing on-target activity and three off-target (OT) loci that, importantly, were at least 10 kb from a coding sequence. This study provides proof-of-concept for TALEN-mediated in situ correction of an endogenous patient-specific gene mutation and used an unbiased screen for comprehensive TALEN target mapping that will cooperatively facilitate translational application.
Sigma-Aldrich
PKH26 Reference Microbeads
Pricing and availability is not currently available.