Merck
  • Optimization of the Linker Domain in a Dimeric Compound that Degrades an r(CUG) Repeat Expansion in Cells.
Optimization of the Linker Domain in a Dimeric Compound that Degrades an r(CUG) Repeat Expansion in Cells.
Journal of medicinal chemistry (2020-07-14)
Raphael I Benhamou, Masahito Abe, Shruti Choudhary, Samantha M Meyer, Alicia J Angelbello, Matthew D Disney
ABSTRACT
RNA repeat expansions are responsible for more than 30 incurable diseases. Among them is myotonic dystrophy type 1 (DM1), the most common form of adult on-set muscular dystrophy. DM1 is caused by an r(CUG) repeat expansion [r(CUG)exp] located in the 3' untranslated region (UTR) of the dystrophia myotonica protein kinase gene. This repeat expansion is highly structured, forming a periodic array of 5'CUG/3'GUC internal loop motifs. We therefore designed dimeric compounds that simultaneously bind two of these motifs by connecting two RNA-binding modules with peptoid linkers of different geometries and lengths. The optimal linker contains two proline residues and enhances compound affinity. Equipping this molecule with a bleomycin A5 cleaving module converts the simple binding compound into a potent allele-selective cleaver of r(CUG)exp. This study shows that the linker in modularly assembled ligands targeting RNA can be optimized to afford potent biological activity.
Sigma-Aldrich
Anti-MBNL1 Antibody, clone 4A8, clone 4A8, from mouse