Merck
  • BMP4 overexpression induces the upregulation of APP/Tau and memory deficits in Alzheimer's disease.
BMP4 overexpression induces the upregulation of APP/Tau and memory deficits in Alzheimer's disease.
Cell death discovery (2021-03-17)
Xiaoqing Zhang, Juan Li, Li Ma, Hui Xu, Yun Cao, Wei Liang, Jia Ma, Z Peter Wang, Yuyun Li
ABSTRACT
Alzheimer's disease (AD) is a chronic progressive degenerative disease of the nervous system. Its pathogenesis is complex and is related to the abnormal expression of the amyloid β (Aβ), APP, and Tau proteins. Evidence has demonstrated that bone morphogenetic protein 4 (BMP4) is highly expressed in transgenic mouse models of AD and that endogenous levels of BMP4 mainly affect hippocampal function. To determine whether BMP4 participates in AD development, transgenic mice were constructed that overexpress BMP4 under the control of the neuron-specific enolase (NSE) promoter. We also performed MTT, FACS, transfection, TUNEL, and Western blotting assays to define the role of BMP4 in cells. We found that middle-aged BMP4 transgenic mice exhibited impaired memory via the Morris water maze experiment. Moreover, their hippocampal tissues exhibited high expression levels of AD-related proteins, including APP, Aβ, PSEN-1, Tau, P-Tau (Thr181), and P-Tau (Thr231). Furthermore, in multiple cell lines, the overexpression of BMP4 increased the expression of AD-related proteins, whereas the downregulation of BMP4 demonstrated opposing effects. Consistent with these results, BMP4 modulation affected cell apoptosis via the regulation of BAX and Bcl-2 expression in cells. Our findings indicate that BMP4 overexpression might be a potential factor to induce AD.
Sigma-Aldrich
MISSION® esiRNA, targeting human BMP4