Merck
  • 2-Substituted (S)-2-(3,3-dimethyl-1-oxo-10,10a-dihydroimidazo[1,5-b]isoquinolin-2(1H,3H,5H)-yl)acetic acids: Conformational prediction, synthesis, anti-thrombotic and vasodilative evaluation.
2-Substituted (S)-2-(3,3-dimethyl-1-oxo-10,10a-dihydroimidazo[1,5-b]isoquinolin-2(1H,3H,5H)-yl)acetic acids: Conformational prediction, synthesis, anti-thrombotic and vasodilative evaluation.
Bioorganic & medicinal chemistry (2011-01-05)
Hong Wang, Le Peng, Ming Zhao, Jiawang Liu, Xiaoyi Zhang, Yuji Wang, Jianhui Wu, Li Li, Shiqi Peng
ABSTRACT
(S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid (TIC) can inhibit thrombosis by inhibiting platelet aggregation. The investigation of amino acids modified TIC reveals that a stretching conformation is critical for high anti-thrombotic activity. The conformational modeling shows that introducing a ring into amino acid modified TIC results in a desirable stretching conformation. According to this hypothesis, we synthesized seventeen novel 2-substituted (S)-2-(3,3-dimethyl-1-oxo-10,10a-dihydroimidazo[1,5-b]isoquinolin-2(1H,3H,5H)-yl)acetic acids (5a-q). In the in vitro anti-platelet aggregation assay, for ADP-induced platelet aggregation the IC(50) values of 5a-q are 1.8-3.4-folds lower than that of TIC. In the in vivo anti-thrombotic assay, the effective dose of 5a-q was 167-folds lower than that of TIC. The vessel strip assay showed that 5a-q had mild vasorelaxation activity.
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%