Merck
  • Synthetic Transition from Thiourea-Based Compounds to Tetrazole Derivatives: Structure and Biological Evaluation of Synthesized New N-(Furan-2-ylmethyl)-1H-tetrazol-5-amine Derivatives.
Synthetic Transition from Thiourea-Based Compounds to Tetrazole Derivatives: Structure and Biological Evaluation of Synthesized New N-(Furan-2-ylmethyl)-1H-tetrazol-5-amine Derivatives.
Molecules (Basel, Switzerland) (2021-01-14)
Daniel Szulczyk, Anna Bielenica, Piotr Roszkowski, Michał A Dobrowolski, Wioletta Olejarz, Sebastian Kmiecik, Małgorzata Podsiad, Marta Struga
ABSTRACT
Twelve novel derivatives of N-(furan-2-ylmethyl)-1H-tetrazol-5-amine were synthesized. For obtained compound 8, its corresponding substrate single crystals were isolated and X-ray diffraction experiments were completed. In the initial stage of research, in silico structure-based pharmacological prediction was conducted. All compounds were screened for their antibacterial and antimycobacterial activities using standard and clinical strains. The cytotoxic activity was evaluated against a panel of human cancer cell lines, in contrast to normal (HaCaT) cell lines, by using the MTT method. All examined derivatives were found to be noncytotoxic against normal cell lines. Within the studied group, compound 6 showed the most promising results in antimicrobial studies. It inhibited four hospital S. epidermidis rods' growth, when applied at the amount of 4 µg/mL. However, the most susceptible to the presence of compound 6 was S. epidermidis T 5501 851/19 clinical strain, for which the MIC value was only 2 µg/mL. Finally, a pharmacophore model was established based on lead compounds from this and our previous work.
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%