Merck
  • Systematic comparison of the structure of homoleptic tetradentate N2O2-type Schiff base complexes of tetravalent f-elements (M(IV) = Ce, Th, U, Np, and Pu) in solid state and in solution.
Systematic comparison of the structure of homoleptic tetradentate N2O2-type Schiff base complexes of tetravalent f-elements (M(IV) = Ce, Th, U, Np, and Pu) in solid state and in solution.
Dalton transactions (Cambridge, England : 2003) (2020-11-21)
Thomas Radoske, Roger Kloditz, Sebastian Fichter, Juliane März, Peter Kaden, Michael Patzschke, Moritz Schmidt, Thorsten Stumpf, Olaf Walter, Atsushi Ikeda-Ohno
ABSTRACT
A series of tetradentate N2O2-type Schiff base complexes with tetravalent 4f- and 5f-block metals, [M(salpn)2] (H2salpn = N,N'-disalicylidene-1,3-diaminopropane; M = Ce, Th, U, Np, and Pu), were prepared to systematically investigate their solid state structure, and their complexation behaviour in solution with the goal to investigate the subtle differences between 4f- and 5f-elements. X-ray diffraction revealed that all investigated metal cations form [M(salpn)2] complexes. All the complexes show the same ligand arrangement with meridional conformation, amongst which only Ce(iv) exhibits unique behaviour upon crystallisation. [Ce(salpn)2] crystallises in two less symmetric systems (P1[combining macron] or P21/n), whilst all the other [M(salpn)2] crystallise in a more symmetric orthorhombic system (Pban). Quantum chemical calculations suggest that the observed structural peculiarity of Ce(iv) stems from the geometrical flexibility due to the more "ionic" nature of bonds to the 4f element. 1H NMR measurements revealed that [M(salpn)2] forms two different species in solution with and without an additional solvent molecule, where the relative distribution of the two species depends mainly on the ionic radius of the metal centre. Again, Ce(iv) behaves differently from the tetravalent actinides with a higher ratio of the solvent-molecule-coordinated species than the ratio expected from its ionic radius. Hence, this study is successful in observing subtle differences between 4f- (i.e. Ce) and 5f-elements (actinides; Th, U, Np, and Pu) both in the solid state and in solution on an analytically distinguishable level, and in relating the observed subtle differences to their electronic structure.
Sigma-Aldrich
N,N′-Bis(salicylidene)-1,3-propanediamine, 99%
Pricing and availability is not currently available.