Merck
  • Emergence of metallo-β-lactamase NDM-1-producing multidrug-resistant Escherichia coli in Australia.
Emergence of metallo-β-lactamase NDM-1-producing multidrug-resistant Escherichia coli in Australia.
Antimicrobial agents and chemotherapy (2010-09-09)
Laurent Poirel, Emilie Lagrutta, Peter Taylor, Jeanette Pham, Patrice Nordmann
ABSTRACT
A multidrug-resistant Escherichia coli isolate recovered in Australia produced a carbapenem-hydrolyzing β-lactamase. Molecular investigations revealed the first identification of the bla(NDM-1) metallo-β-lactamase gene in that country. In addition, this E. coli isolate expressed the extended-spectrum β-lactamase CTX-M-15, together with two 16S rRNA methylases, namely, ArmA and RmtB, conferring a high level of resistance to aminoglycosides.
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Pricing and availability is not currently available.