Merck
  • Synthesis and evaluation of phenylequine for antimalarial activity in vitro and in vivo.
Synthesis and evaluation of phenylequine for antimalarial activity in vitro and in vivo.
Bioorganic & medicinal chemistry letters (2009-12-26)
Margaret A L Blackie, Vanessa Yardley, Kelly Chibale
ABSTRACT
Synthesis of the potent antiplasmodial 4-aminoquinoline, phenylequine (PQ), is reported for the first time. PQ and the two analogues show increased efficacy in moving from the chloroquine sensitive D10 to the chloroquine resistant K1 strain in vitro. The in vivo efficacy of PQ, and salts thereof, have been determined in Plasmodium berghei ANKA and Plasmodium yoelii. Phenylequine hydrochloride has shown an ED(50) of 0.81 in P. yoelii (cf chloroquine ED(50)=1.31).
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)